4-Chlorobenzyl alcohol

Supplier: Thermo Scientific

Synonyms: (4-Chlorophenyl)methanol

SB01197.1 SB01197EA SB01197EE
MAYBSB01197.1EA 0 EUR
MAYBSB01197.1 MAYBSB01197EA MAYBSB01197EE
4-Chlorobenzyl alcohol
4-Chlorobenzyl alcohol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 67…70 °C
MDL Number: MFCD00004652
CAS Number: 873-76-7
EINECS: 212-852-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR