Benzyl cyanide, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

B19401-1KG B19401-250G
SIALB19401-1KGEA 106 EUR
SIALB19401-1KG SIALB19401-250G
Benzyl cyanide, Sigma-Aldrich®
Benzyl cyanide
Formula: (C₆H₅)CH₂CN
MW: 117.15 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00001894
CAS Number: 140-29-4
EINECS: 205-410-5
UN: 2470
ADR: 6.1,III
Merck Index: 13,01133

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR