5-Methylisophthalonitrile
Supplier: Thermo Scientific
|
Warning
|
CD03148.1
CD03148.10
MAYBCD03148.1EA
0
EUR
MAYBCD03148.1
MAYBCD03148.10
5-Methylisophthalonitrile
5-Methylisophthalonitrile
Formula:
C₉H₆N₂ Boiling Pt: 311 °C (1013 hPa) Melting Pt: 163.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052016 CAS Number: 39718-07-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...