Butyrophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Phenylbutan-1-one, Phenyl propyl ketone, 1-Phenyl-1-butanone
M-Clarity™ quality level = MQ100
124338-100G 124338-25G
SIAL124338-100GEA 189 EUR
SIAL124338-100G SIAL124338-25G
Butyrophenone, Sigma-Aldrich®
Butyrophenone
Formula: C₆H₅C(O)(CH₂)₂CH₃
MW: 148.2 g/mol
Boiling Pt: 220 °C (1013 hPa)
Density: 0.988 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009397
CAS Number: 495-40-9
EINECS: 207-799-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d21a84719c71ec28f51844913f7376027ac053&vid=a8921a2a2e416872169c2c45fe321889cd871e13&prid=web&clid=SAPDAM