(1-Bromoethyl)benzene ≥97%, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Bromo-1-phenylethane, α-Methylbenzyl bromide
M-Clarity™ quality level = MQ100
238104-25G 238104-100G
SIAL238104-25GEA 57.6 EUR
SIAL238104-25G SIAL238104-100G
(1-Bromoethyl)benzene ≥97%, Sigma-Aldrich®
1-Bromo-1-phenylethane
Formula: C₆H₅CHBrCH₃
MW: 185.06 g/mol
Boiling Pt: 94 °C (16 mmHg) (lit.)
Melting Pt: 50
Density: 1.356 g/mL (25 °C) (lit.)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000139
CAS Number: 585-71-7
EINECS: 209-560-2
UN: 3334
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f329f7855ae4dffcd097f49d6d504b1ec165e692&vid=6d612184698aeb1969948bdeb536d2ee39ec9551&prid=web&clid=SAPDAM