1-Phenyl-2-butanone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Phenylbutan-2-one, Benzyl ethyl ketone
137227-25G
137227-5G
SIAL137227-25GEA
549
EUR
SIAL137227-25G
SIAL137227-5G
1-Phenyl-2-butanone, Sigma-Aldrich®
1-Phenyl-2-butanone
Formula:
C₆H₅C(O)CH₂CH₂CH₃ MW: 148.2 g/mol Boiling Pt: 230 °C (1013 hPa) Melting Pt: 5 °C Density: 0.989 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009315 CAS Number: 1007-32-5 EINECS: 213-752-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...