Benzylacetone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Phenyl-2-butanone, 4-Phenylbutan-2-one

M-Clarity™ quality level = MQ200

B16003-100G B16003-5G
SIALB16003-100GEA 44.2 EUR
SIALB16003-100G SIALB16003-5G
Benzylacetone, Sigma-Aldrich®
Benzylacetone
Formula: (C₆H₅)CH₂CH₂C(O)CH₃
MW: 148.2 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: –13 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00008790
CAS Number: 2550-26-7
EINECS: 219-847-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR