Acetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Methyl phenyl ketone, Phenyl methyl ketone
MQ100
A10701-100ML A10701-1L
SIALA10701-100MLEA 25.4 EUR
SIALA10701-100ML SIALA10701-1L
Acetophenone, Sigma-Aldrich®
Acetophenone
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 201.7
Melting Pt: 20
Density: 1.027 g/cm³ (20 °C)
Flash Pt: 82
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
UN: 3334
ADR: 9
Merck Index: 13,00074