Acetophenone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Methyl phenyl ketone, Phenyl methyl ketone

M-Clarity™ quality level = MQ300

W200905-10KG-K W200905-1KG-K W200905-25KG-K
SIALW200905-10KG-KEA 372 EUR
SIALW200905-10KG-K SIALW200905-1KG-K SIALW200905-25KG-K
Acetophenone, Sigma-Aldrich®
Acetophenone
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 201.7
Melting Pt: 20
Density: 1.027 g/cm³ (20 °C)
Flash Pt: 82
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
UN: 3334
ADR: 9
Merck Index: 13,00074

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR