2'-Aminoacetophenone, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-(2-Aminophenyl)ethanone, o-Aminoacetophenone
M-Clarity™ quality level = MQ100
A37804-25G
A37804-5G
SIALA37804-25GEA
84.8
EUR
SIALA37804-25G
SIALA37804-5G
2'-Aminoacetophenone, Sigma-Aldrich®
2'-Aminoacetophenone
Formula:
C₈H₉NO MW: 135.17 g/mol Boiling Pt: 252 °C (1013 hPa) Melting Pt: 20 °C Density: 1.12 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007717 CAS Number: 551-93-9 EINECS: 209-002-8 Merck Index: 13,00412 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...