4'-Aminoacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-(4-Aminophenyl)ethanone, p-Aminoacetophenone
M-Clarity™ quality level = MQ200
A38002-100G A38002-25G
SIALA38002-100GEA 98.3 EUR
SIALA38002-100G SIALA38002-25G
4'-Aminoacetophenone, Sigma-Aldrich®
4'-Aminoacetophenone
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 101…103 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007896
CAS Number: 99-92-3
EINECS: 202-801-2
Merck Index: 13,00412