2'-Nitroacetophenone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone

M-Clarity™ quality level = MQ100

N9209-10G N9209-25G
SIALN9209-10GEA 28.3 EUR
SIALN9209-10G SIALN9209-25G
2'-Nitroacetophenone, Sigma-Aldrich®
2'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 23 °C
Density: 1.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS Number: 577-59-3
EINECS: 209-414-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR