3'-Nitroacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: m-Nitroacetophenone
N9381-100G N9381-500G
SIALN9381-100GEA 29.7 EUR
SIALN9381-100G SIALN9381-500G
3'-Nitroacetophenone, Sigma-Aldrich®
3'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 77…78 °C
Density: 1.433 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007259
CAS Number: 121-89-1
EINECS: 204-504-3