3'-Methoxyacetophenone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3′-Methoxyacetophenone

M-Clarity™ quality level = MQ100

M9408-100G M9408-25G
SIALM9408-100GEA 152 EUR
SIALM9408-100G SIALM9408-25G
3'-Methoxyacetophenone, Sigma-Aldrich®
3'-Methoxyacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 241
Melting Pt: –8 °C
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008736
CAS Number: 586-37-8
EINECS: 209-573-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR