4'-Methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Acetanisole, 4-Methoxyacetophenone
M-Clarity™ quality level = MQ300
W200506-1KG-K W200506-5KG-K
SIALW200506-1KG-KEA 130 EUR
SIALW200506-1KG-K SIALW200506-5KG-K
4'-Methoxyacetophenone, Sigma-Aldrich®
4'-Methoxyacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 152
Melting Pt: 36…38 °C
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008745
CAS Number: 100-06-1
EINECS: 202-815-9