4'-Methoxypropiophenone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
M24809-25G M24809-5G
SIALM24809-25GEA 69.1 EUR
SIALM24809-25G SIALM24809-5G
4'-Methoxypropiophenone, Sigma-Aldrich®
4'-Methoxypropiophenone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: ∼273 °C (1013 hPa)
Melting Pt: ∼37.5 °C
Density: ∼0.94 g/cm³ (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00009310
CAS Number: 121-97-1
EINECS: 204-512-7