2',4'-Dimethoxyacetophenone ≥98%
Supplier: Thermo Scientific
Synonyms: 1-(2,4-Dimethoxyphenyl)ethan-1-one
A15982.14 A15982.22
A15982.14EA 73.5 EUR
A15982.14 A15982.22
2',4'-Dimethoxyacetophenone ≥98%
2',4'-Dimethoxyacetophenone
Purity: 98% CAS No.: 829-20-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Melting Point: 39-41° Boiling Point: 287-289° MDL No.: MFCD00008727
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 287…289 °C
Melting Pt: 39…41 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008727
CAS Number: 829-20-9
EINECS: 212-587-2