4-Hydroxybenzylacetone, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Raspberry ketone, 1-(4-Hydroxyphenyl)-3-butanone, 4-(4-Hydroxyphenyl)-2-butanone
M-Clarity™ quality level = MQ100
178519-250G
178519-50G
SIAL178519-250GEA
273
EUR
SIAL178519-250G
SIAL178519-50G
4-Hydroxybenzylacetone, Sigma-Aldrich®
4-Hydroxybenzylacetone
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 292 °C (1013 hPa) Melting Pt: 83…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002394 CAS Number: 5471-51-2 EINECS: 226-806-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...