4-Hydroxybenzylacetone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Raspberry ketone, 1-(4-Hydroxyphenyl)-3-butanone, 4-(4-Hydroxyphenyl)-2-butanone
M-Clarity™ quality level = MQ300
W258806-1KG-K W258806-250G-K W258806-5KG-K
SIALW258806-1KG-KEA 127 EUR
SIALW258806-1KG-K SIALW258806-250G-K SIALW258806-5KG-K
4-Hydroxybenzylacetone, Sigma-Aldrich®
4-Hydroxybenzylacetone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 83…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00002394
CAS Number: 5471-51-2
EINECS: 226-806-4