3'-Hydroxyacetophenone 99+%
Supplier: Thermo Fisher Scientific
Warning
411910050 411911000
ACRO411910050EA 19.3 EUR
ACRO411910050 ACRO411911000
3'-Hydroxyacetophenone 99+%
3'-Hydroxyacetophenone
Formula: C₈H₈O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 94…97 °C
Density: 1,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0
Order Now
Specification Test Results
Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 97 °C
GC ≥99.0 %