3'-Hydroxyacetophenone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
H18801-100G H18801-25G
SIALH18801-100GEA 76.1 EUR
SIALH18801-100G SIALH18801-25G
3'-Hydroxyacetophenone, Sigma-Aldrich®
3'-Hydroxyacetophenone
Formula: C₈H₈O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 86…88 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0