2',4'-Dihydroxypropiophenone ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,4-Dihydroxypropiophenone
B23414.14
B23414.22
B23414.14EA
69.6
EUR
B23414.14
B23414.22
2',4'-Dihydroxypropiophenone ≥99%
2',4'-Dihydroxypropiophenone
Purity: 99%
CAS No.: 5792-36-9
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 98-101°
Beilstein Reference: 1238635
Formula:
C₉H₁₀O₃ Boiling Pt: 343 °C (1013 hPa) Melting Pt: 98…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002280 CAS Number: 5792-36-9 EINECS: 227-329-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...