3',5'-Dihydroxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone
M-Clarity™ quality level = MQ100
224596-10G
224596-50G
SIAL224596-10GEA
159
EUR
SIAL224596-10G
SIAL224596-50G
3',5'-Dihydroxyacetophenone, Sigma-Aldrich®
3',5'-Dihydroxyacetophenone
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 148 °C |
MDL Number:
MFCD00002290 CAS Number: 51863-60-6 EINECS: 257-480-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...