3-Toluoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 3-Methylbenzoyl chloride, m-Methylbenzoyl chloride, m-Toluoyl chloride, 3-Methylbenzene-1-carbonyl chloride

M-Clarity™ quality level = MQ100

122254-100G 122254-5G
SIAL122254-100GEA 93 EUR
SIAL122254-100G SIAL122254-5G
3-Toluoyl chloride, Sigma-Aldrich®
3-Toluoyl chloride
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: –23 °C
Density: 1.172 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000681
CAS Number: 1711-06-4
EINECS: 216-976-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR