4-tert-Butylbenzoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 4-tert-Butylbenzoylchloride

M-Clarity™ quality level = MQ100

157120-100G 157120-25G
SIAL157120-100GEA 222 EUR
SIAL157120-100G SIAL157120-25G
4-tert-Butylbenzoyl chloride, Sigma-Aldrich®
4-tert-Butylbenzoyl chloride
Formula: C₁₁H₁₃ClO
MW: 196.68 g/mol
Boiling Pt: 135 °C (1013 hPa)
Density: 1.0864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000695
CAS Number: 1710-98-1
EINECS: 216-973-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR