4-Butylbenzoyl chloride

Supplier: Thermo Scientific

BTB09878.1 BTB09878.10 BTB09878.50
MAYBBTB09878.1EA 0 EUR
MAYBBTB09878.1 MAYBBTB09878.10 MAYBBTB09878.50
4-Butylbenzoyl chloride
4-Butylbenzoyl chloride
Formula: C₁₁H₁₃ClO
MW: 196.68 g/mol
Boiling Pt: 155 °C (1013 hPa)
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00000699
CAS Number: 28788-62-7
EINECS: 249-224-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR