4-Toluoyl chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
p-Toluoyl chloride, 4-Methylbenzene-1-carbonyl chloride, p-Methylbenzoyl chloride, 4-Methylbenzoyl chloride
M-Clarity™ quality level = MQ100
106631-100G
106631-500G
106631-5G
SIAL106631-100GEA
58.7
EUR
SIAL106631-100G
SIAL106631-500G
SIAL106631-5G
4-Toluoyl chloride, Sigma-Aldrich®
4-Toluoyl chloride
Formula:
C₈H₇ClO Boiling Pt: 225 °C (1013 hPa) Melting Pt: –2 °C Density: 1.175 g/cm³ (20 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000696 CAS Number: 874-60-2 EINECS: 212-864-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...