o-Toluic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Methylbenzoic acid
T36404-100G T36404-500G
SIALT36404-100GEA 52.2 EUR
SIALT36404-100G SIALT36404-500G
o-Toluic acid, Sigma-Aldrich®
o-Toluic acid
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 101…104 °C
Density: 1.2555 g/cm³ (20 °C)
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002477
CAS Number: 118-90-1
EINECS: 204-284-9
Merck Index: 12,09673