4-tert-Butylbenzoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-(tert-Butyl)benzoic acid
M-Clarity™ quality level = MQ100
150355-2KG 150355-500G
SIAL150355-2KGEA 136 EUR
SIAL150355-2KG SIAL150355-500G
4-tert-Butylbenzoic acid, Sigma-Aldrich®
4-tert-Butylbenzoic acid
Formula: C₁₁H₁₄O₂
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 165…167 °C
Density: 0.6 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002563
CAS Number: 98-73-7
EINECS: 202-696-3
UN: 2811
ADR: 6.1,III