4-Isopropylbenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
268402-25G
268402-5G
SIAL268402-25GEA
125
EUR
SIAL268402-25G
SIAL268402-5G
4-Isopropylbenzoic acid, Sigma-Aldrich®
4-Isopropylbenzoic acid
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 115…117 °C Density: 1.162 g/cm³ (20 °C) |
MDL Number:
MFCD00002564 CAS Number: 536-66-3 EINECS: 208-642-5 Merck Index: 12,02684 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...