p-Toluic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Methylbenzoic acid, 4-Toluic acid
M-Clarity™ quality level = MQ100
T36803-100G T36803-500G
SIALT36803-100GEA 51.6 EUR
SIALT36803-100G SIALT36803-500G
p-Toluic acid, Sigma-Aldrich®
p-Toluic acid
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 179…180 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002565
CAS Number: 99-94-5
EINECS: 202-803-3
Merck Index: 12,09673