5-Methylanthranil acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid
M-Clarity™ quality level = MQ100
419443-25G 419443-5G
SIAL419443-25GEA 389 EUR
SIAL419443-25G SIAL419443-5G
5-Methylanthranil acid, Sigma-Aldrich®
5-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 317
Storage Temperature: Ambient
MDL Number: MFCD00007909
CAS Number: 2941-78-8
EINECS: 220-932-3