6-Nitro-m-toluic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid

M-Clarity™ quality level = MQ100

113409-25G 113409-5G
SIAL113409-25GEA 140 EUR
SIAL113409-25G SIAL113409-5G
6-Nitro-m-toluic acid, Sigma-Aldrich®
6-Nitro-m-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: 137…138 °C
MDL Number: MFCD00007368
CAS Number: 3113-72-2
EINECS: 221-481-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR