Benzotrifluoride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: α,α,α-Trifluorotoluene, Solvent BTF

M-Clarity™ quality level = MQ100

547948-100ML 547948-1L 547948-2L
SIAL547948-100MLEA 32.5 EUR
SIAL547948-100ML SIAL547948-1L SIAL547948-2L
Benzotrifluoride, Sigma-Aldrich®
Benzotrifluoride
Formula: C₇H₅F₃
MW: 146.11 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.37 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000372
CAS Number: 98-08-8
EINECS: 202-635-0
UN: 2338
ADR: 3,II
Merck Index: 13,01111
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05485864abd8b35273efdbb2e08a5ed79b907869&vid=3bc01224d36fbaaa4afb501b0dd876d8c9eaf997&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR