α,α,α,4-Tetrafluorotoluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Fluorobenzotrifluoride, 1-Fluoro-4-(trifluoromethyl)benzene
M-Clarity™ quality level = MQ100
195359-25G 195359-5G
SIAL195359-25GEA 91.3 EUR
SIAL195359-25G SIAL195359-5G
α,α,α,4-Tetrafluorotoluene, Sigma-Aldrich®
α,α,α,4-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 102 °C (1013 hPa)
Density: 1.293 g/cm³ (25 °C)
MDL Number: MFCD00000399
CAS Number: 402-44-8
EINECS: 206-944-1
UN: 1993
ADR: 3,II