(2-Fluorophenyl)acetonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Fluorobenzyl cyanide, 2-(2-Fluorophenyl)acetonitrile
M-Clarity™ quality level = MQ100
199346-10G 199346-50G
SIAL199346-10GEA 45 EUR
SIAL199346-10G SIAL199346-50G
(2-Fluorophenyl)acetonitrile, Sigma-Aldrich®
(2-Fluorophenyl)acetonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 1.14045 g/cm³ (15 °C)
MDL Number: MFCD00001897
CAS Number: 326-62-5
EINECS: 206-313-0
UN: 3276
ADR: 6.1,III