4'-Fluoroacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
M-Clarity™ quality level = MQ100
F3207-100G F3207-25G
SIALF3207-100GEA 89.3 EUR
SIALF3207-100G SIALF3207-25G
4'-Fluoroacetophenone, Sigma-Aldrich®
4'-Fluoroacetophenone
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 196
Density: 1.138 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9