α,α,α-Trifluoro-p-toluoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 4-(Trifluoromethyl)benzoyl chloride, 4-(Trifluoromethyl)benzoylchloride, 4-(Trifluoromethyl)benzene-1-carbonyl chloride

M-Clarity™ quality level = MQ100

249475-1G 249475-25G 249475-5G
SIAL249475-1GEA 34.4 EUR
SIAL249475-1G SIAL249475-25G SIAL249475-5G
α,α,α-Trifluoro-p-toluoyl chloride, Sigma-Aldrich®
α,α,α-Trifluoro-p-toluoyl chloride
Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
Boiling Pt: 188…190 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.4035 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000694
CAS Number: 329-15-7
EINECS: 206-342-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR