α,α,α-Trifluoro-o-toluic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-(Trifluoromethyl)benzoic acid
188450050 188450250
ACRO188450050EA 35.4 EUR
ACRO188450050 ACRO188450250
α,α,α-Trifluoro-o-toluic acid 98%
α,α,α-Trifluoro-o-toluic acid
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 247 °C (753 mmHg)
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002476
CAS Number: 433-97-6
EINECS: 207-093-9
Order Now
Specification Test Results
Appearance White to yellow-brown Powder
Infrared spectrum Conforms
Melting point 106 to 112 °C
HPLC ≥97.5 %