α,α,α-Trifluoro-o-toluic acid

Supplier: Thermo Scientific
Warning

Synonyms: 2-(Trifluoromethyl)benzoic acid

SB01835.1 SB01835.10
MAYBSB01835.1EA 0 EUR
MAYBSB01835.1 MAYBSB01835.10
α,α,α-Trifluoro-o-toluic acid
α,α,α-Trifluoro-o-toluic acid
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 110 °C
MDL Number: MFCD00002476
CAS Number: 433-97-6
EINECS: 207-093-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR