3-Fluoro-o-toluic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

342114-1G 342114-5G
SIAL342114-1GEA 27.8 EUR
SIAL342114-1G SIAL342114-5G
3-Fluoro-o-toluic acid, Sigma-Aldrich®
3-Fluoro-o-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00075246
CAS Number: 699-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR