α,α,α,5-Tetrafluoro-m-toluic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Fluoro-5-trifluoromethylbenzoic acid, 3-Fluoro-5-(trifluoromethyl)benzoic acid
M-Clarity™ quality level = MQ100
455326-1G
455326-5G
SIAL455326-1GEA
38
EUR
SIAL455326-1G
SIAL455326-5G
α,α,α,5-Tetrafluoro-m-toluic acid, Sigma-Aldrich®
α,α,α,5-Tetrafluoro-m-toluic acid
Formula:
C₈H₄F₄O₂ MW: 208.11 g/mol |
MDL Number:
MFCD00061293 CAS Number: 161622-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...