α,α,α-Trifluoro-p-toluic acid
Supplier: Thermo Scientific
Warning
Synonyms: 4-(Trifluoromethyl)benzoic acid
SB00672.1 SB00672.10
MAYBSB00672.1EA 0 EUR
MAYBSB00672.1 MAYBSB00672.10
α,α,α-Trifluoro-p-toluic acid
α,α,α-Trifluoro-p-toluic acid
Formula: C₈H₅F₃O₂
Melting Pt: 219…220 °C
MDL Number: MFCD00002562
CAS Number: 455-24-3
EINECS: 207-242-8