2-Iodo-4-(trifluoromethyl)aniline ≥99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Amino-3-iodobenzotrifluoride, 2-Iodo-α,α,α-trifluoro-p-toluidine, 4-Amino-3-iodo-α,α,α-trifluorotoluene
B22691.09 B22691.18 B22691.30 AB131379.0250
B22691.09EA 71.7 EUR
B22691.09 B22691.18 B22691.30 ABCRAB131379.0250
2-Iodo-4-(trifluoromethyl)aniline ≥99%
2-Iodo-4-(trifluoromethyl)aniline
Purity: 99% CAS No.: 163444-17-5 Molecular Formula: C7H5F3IN Formula Weight: 287.02 Melting Point: 49-52° MDL No.: MFCD02683097
Formula: C₇H₅F₃IN
MW: 287.02 g/mol
Melting Pt: 49…52 °C
Storage Temperature: Ambient
MDL Number: MFCD02683097
CAS Number: 163444-17-5