4-Fluorobenzylamine 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (4-Fluorophenyl)methanamine
171950250 171951000
ACRO171950250EA 56.6 EUR
ACRO171950250 ACRO171951000
4-Fluorobenzylamine 97%
4-Fluorobenzylamine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 183…185 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008120
CAS Number: 140-75-0
EINECS: 205-430-4
UN: 2735
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5115 to 1.5155 (20°C, 589 nm)
GC ≥96.0 %