4-Fluorobenzylamine
Supplier: Thermo Scientific
Danger
Synonyms: (4-Fluorophenyl)methanamine
SB01159.1 SB01159.50
MAYBSB01159.1EA 0 EUR
MAYBSB01159.1 MAYBSB01159.50
4-Fluorobenzylamine
4-Fluorobenzylamine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 183
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008120
CAS Number: 140-75-0
EINECS: 205-430-4
UN: 2735
ADR: 8 III