4-Fluorobenzylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
(4-Fluorophenyl)methanamine
M-Clarity™ quality level = MQ100
162493-25G
162493-5G
SIAL162493-25GEA
61.6
EUR
SIAL162493-25G
SIAL162493-5G
4-Fluorobenzylamine, Sigma-Aldrich®
4-Fluorobenzylamine
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 183 °C (1013 hPa) Density: 1.09 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008120 CAS Number: 140-75-0 EINECS: 205-430-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...