5-Fluoro-o-toluidine
Supplier: Thermo Scientific
Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline
SB00690.1 SB00690.10 SB00690.50
MAYBSB00690.1EA 0 EUR
MAYBSB00690.1 MAYBSB00690.10 MAYBSB00690.50
5-Fluoro-o-toluidine
5-Fluoro-o-toluidine
Formula: C₇H₈FN
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 36…38 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007764
CAS Number: 367-29-3
EINECS: 206-689-6
UN: 2811
ADR: 6.1,III