4-Fluoro-3-(trifluoromethyl)aniline

Supplier: Thermo Scientific
Warning

Synonyms: 5-Amino-2-fluorobenzotrifluoride, 5-Amino-α,α,α,2-tetrafluorotoluene, α,α,α,4-Tetrafluoro-m-toluidine, 5-Amino-alpha,alpha,alpha,2-tetrafluorotoluene

SB01727.1 SB01727.10 SB01727.50
MAYBSB01727.1EA 0 EUR
MAYBSB01727.1 MAYBSB01727.10 MAYBSB01727.50
4-Fluoro-3-(trifluoromethyl)aniline
4-Fluoro-3-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 207 °C (1013 hPa)
Density: 1.39 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007834
CAS Number: 2357-47-3
EINECS: 219-095-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR