2-Fluoro-5-nitrobenzotrifluoride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene
M-Clarity™ quality level = MQ100
365823-25G 365823-5G
SIAL365823-25GEA 66.3 EUR
SIAL365823-25G SIAL365823-5G
2-Fluoro-5-nitrobenzotrifluoride, Sigma-Aldrich®
2-Fluoro-5-nitrobenzotrifluoride
Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Boiling Pt: 99…110 °C (35 mmHg)
Melting Pt: 22…24 °C
Density: 1.52 g/cm³ (20 °C)
MDL Number: MFCD00024576
CAS Number: 400-74-8
EINECS: 206-925-8
UN: 2810
ADR: 6.1,III